Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Thermoset F217 and PTFE Caps, Cleaned for Volatiles

Catalog No. 02992412
Change view
Click to view available options
Capacity (Metric):
60 mL
120 mL
240 mL
Closure Size:
53-400 mm
58-400 mm
70-400 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
02-992-412 60 mL 53-400 mm
02-992-399 120 mL 58-400 mm
02-992-390 240 mL 70-400 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-992-412 Supplier Qorpak™ Supplier No. GLC01612

missing translation for 'validMainframeMsg'

Offers maximum content visibility and integrity

  • Without shoulders for maximum storage capacity
  • Green cap easily identifies PTFE liner system
  • Usage: analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents

Certifications

Meets U.S. EPA Specifications and Guidelines for Contaminant-Free Sample Containers

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Color Clear
Diameter (English) 2.17 in.
Material Glass
Quantity 24/Cs.
Closure Color Green
Height (Metric) 49 mm
Height (English) 1.93 in.
Closure Liner F217, PTFE
Closure Material Thermoset
Closure Size 53-400 mm
Diameter (Metric) 55 mm
Closure Type Screw Cap
Capacity (English) 2 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.