Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Thermoset F217 and PTFE Lined Caps

Catalog No. 02991752
Change view
Click to view available options
Capacity (Metric):
960 mL
Quantity:
12/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
02-991-752 960 mL 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-752 Supplier Qorpak™ Supplier No. GLC01720

missing translation for 'validMainframeMsg'

Offers maximum content visibility

  • Without shoulders for maximum storage capacity
  • Green cap easily identifies PTFE liner system
  • Usage: analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents

Specifications

Capacity 32 oz., 960mL
Capacity (Metric) 960 mL
Color Clear
Diameter (English) 3.74 in.
Material Glass
Quantity 12/Cs.
Closure Color Green
Height (Metric) 170 mm
Height (English) 6.69 in.
Closure Liner F217, PTFE
Closure Material Thermoset
Closure Size 89-400 mm
Diameter (Metric) 95 mm
Closure Type Screw Cap
Capacity (English) 32 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.