Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ CurTec™ Innnerseals for White Packo Containers

Catalog No. 02994108
Change view
Click to view available options
For Use With (Equipment):
1500 to 2500 mL white pack containers
300 to 1300 mL white pack containers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
02-994-108 300 to 1300 mL white pack containers
02-994-109 1500 to 2500 mL white pack containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-994-108 Supplier Qorpak™ Supplier No. 248746
Add to Cart
Add to Cart

For use with Packo containers. Qorpak™ CurTec™ Innerseals provides high level of product protection and added value through quality, functionality, ease of use, and design.

  • Made of polyethylene
  • Translucent seal

Specifications

Color Natural
Material Polyethylene
Sterility Non-sterile
Liner Material Polyethylene
Disposable Single-use
Product Type Cap Sealing Disc
For Use With (Equipment) 300 to 1300 mL white pack containers
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.