Learn More
Qorpak™ CurTec™ Lids for White Packo Containers
Description
- Made of polypropylene
- Available with and without gasket
Specifications
Specifications
| Color | Dark Red |
| Material | Polypropylene |
| No. per Case | 30/Cs. |
| Packaging | Convenience |
| Sterility | Non-sterile |
| For Use With (Equipment) | 1500 to 2500 mL Packo Containers |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Product Type | Quarter-turn Screw Lid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.