Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ CurTec™ Wide Neck Drums

Catalog No. 02994116
Change view
Click to view available options
Diameter (English) Outer:
7.7 in.
10.7 in.
12.4 in.
Height (English):
9.3 in.
10.3 in.
12.8 in.
16.4 in.
16.7 in.
Weight (English):
30 lb.
32 lb.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Outer Height (English) Weight (English)
02-994-116 10.7 in. 12.8 in. 32 lb.
02-994-115 10.7 in. 9.3 in. 30 lb.
02-994-118 12.4 in. 16.7 in. 32 lb.
02-994-114 7.7 in. 10.3 in. 30 lb.
02-994-117 10.7 in. 16.4 in. 32 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-994-116 Supplier Qorpak™ Supplier No. 248732
Add to Cart
Add to Cart

Provides high level of product protection and added value through quality, functionality, ease of use, and design. Qorpak™ CurTec™ Wide Neck Drums are used for shipping and stocking valuable fine chemicals.

  • Features red polypropylene unlined lid
  • Wide neck for easy filling of contents

Certifications

HACCP, ISO

Specifications

Certifications/Compliance HACCP, ISO
Product Type Drum/Pail with Screw-on Lid
Closure Type Cap
Capacity (English) 507 oz.
Capacity (Metric) 15 L
Color White
For Use With (Application) Ideal for pharmaceuticals, fine chemicals, food ingredients and hazardous waste
Material HDPE
Height (English) 12.8 in.
Height (Metric) 327 mm
Diameter (English) Outer 10.7 in.
Diameter (Metric) Outer 274 mm
Weight (English) 32 lb.
Weight (Metric) 14.5 Kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.