Learn More
Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Specifications
| Capacity | 2 oz., 60 mL |
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Clear |
| Diameter (English) | 1.65 in. |
| Material | Glass |
| Quantity | 48/Cs. |
| Sterility | Non-sterile |
| Closure Color | Green |
| Closure Included | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.