Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Metal Innerseals for F-Style Cans

Catalog No. 02991621
Change view
Click to view available options
Material:
Metal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
02-991-621 Metal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-621 Supplier Qorpak™ Supplier No. MET05442
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For added protection of contents in F-style cans

  • For use with F-style cans

Specifications

Product Type Innerseal Can
Description Alpha
Color Silver
For Use With (Application) For added protection to contents of F Style cans
Material Metal
Quantity 15000 Cs.
Diameter (English) Opening 1.25 in.
Diameter (Metric) Opening 3.17 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.