Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Natural LDPE Plug Caps

Catalog No. 02991782
Change view
Click to view available options
Thread Size (Metric):
AP mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02-991-782 AP mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-782 Supplier Qorpak™ Supplier No. CAP00501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offers protection against airborne contaminants

  • Made of natural LDPE
  • Unlined
  • Dispenses a spray mist or drops
  • Best used with Over Caps
  • Often used for nasal spray bottles

Specifications

Color Natural
Material LDPE
No. per Case 7000 Cs.
Sterility Non-sterile
Thread Size (Metric) AP mm
Neck Finish 18
For Use With (Equipment) Over cap
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Threaded Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.