Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Polypropylene F422 HDPE Foam Lined Caps

Catalog No. 02991047
Change view
Click to view available options
Color:
Black
White
Liner Material:
F422 Foam Polyethylene
F422 HDPE Foam
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Liner Material
02-991-047 White F422 HDPE Foam
02-991-033 Black F422 Foam Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-991-047 Supplier Qorpak™ Supplier No. CAP00379
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides resistance to acids

  • Liner is comprised of a 0.38 in. LDPE core sandwiched between identical layers of 0.002 to 0.003 in. virgin HDPE

Specifications

Color White
Material Polypropylene
No. per Case 1100/Cs.
Sterility Non-sterile
Liner Material F422 HDPE Foam
Neck Finish 38-439
For Use With (Equipment) Bottle
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Ribbed Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.