Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Preassembled Hole Caps with PTFE/Silicone Septa

Catalog No. 02991631
Change view
Click to view available options
Thread Size (Metric):
AP mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02-991-631 AP mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-631 Supplier Qorpak™ Supplier No. CAP07511
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides great resealability

  • Resist coring

Specifications

Color Green
Material Polypropylene
No. per Case 3700 Cs.
Sterility Non-sterile
Thread Size (Metric) AP mm
Neck Finish 24-410
For Use With (Equipment) Bottle
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Threaded Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.