Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Polypropylene Hole Caps with Bonded PTFE/Silicone Septa

Catalog No. 02991198
Change view
Click to view available options
Neck Finish:
28-400
33-400
58-400
No. per Case:
1000 Cs.
120 Cs.
Packaging:
Bagged
Bulk
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02-991-198 33-400 120 Cs. Bagged
02-991-201 28-400 120 Cs. Bagged
02-991-417 33-400 1000 Cs. Bulk
02-991-476 28-400 1000 Cs. Bulk
02-991-583 58-400 1000 Cs. Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-991-198 Supplier Qorpak™ Supplier No. CAP06681
Add to Cart
Add to Cart

No slipping away of septa from cap while injecting

  • Good impact strength with pliability
  • Septa features high resealing and chemical resistant properties

Specifications

Color White
Material Polypropylene
No. per Case 120 Cs.
No. per Pack 12/pack
Packaging Bagged
Sterility Non-sterile
Liner Material PTFE/Silicone
Neck Finish 33-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.