Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Unlined Polypropylene Caps

Catalog No. 02991447
Change view
Click to view available options
Neck Finish:
120-400
33-400
38-400
43-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
1100 Caps/Cs.
1300 Caps/Cs.
144 Caps/Cs.
1700 Caps/Cs.
2200 Caps/Cs.
242 Caps/Cs.
2900 Caps/Cs.
4000 Caps/Cs.
456 Caps/Cs.
760 Caps/Cs.
900 Caps/Cs.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
02-991-447 63-400 900 Caps/Cs.
02-991-444 33-400 4000 Caps/Cs.
02-991-445 38-400 2900 Caps/Cs.
02-991-274 43-400 2200 Caps/Cs.
02-991-144 48-400 1700 Caps/Cs.
02-991-145 53-400 1300 Caps/Cs.
02-991-446 58-400 1100 Caps/Cs.
02-991-345 70-400 760 Caps/Cs.
02-991-122 89-400 456 Caps/Cs.
02-991-109 120-400 242 Caps/Cs.
02-993-135 89-400 144 Caps/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 02-991-447 Supplier Qorpak™ Supplier No. CAP00403
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polypropylene caps are known for good impact strength, durability, cost-effectiveness, thermal resistance, and pliability.

  • Autoclavable
  • Well-suited for many dry products
  • Add a PTFE disc liner to the unlined polypropylene cap for better leak resistance when autoclaving

Specifications

Color White
Material Polypropylene
No. per Case 900 Caps/Cs.
Packaging Bulk
Sterility Non-sterile
Neck Finish 63-400
For Use With (Equipment) Wide Mouth Bottle
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Ribbed Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.