Learn More
Hach Company QP1680 Consumables Kit, 2500 Analysis - For QP1680 TOC/TN Analyzer
Catalog No. NC3893355 Shop All Hach Company Products
Encompass
Consumables kit supplying replacement parts for approximately 2,500 injections on the QP1680 TOC/TN analyzer. The kit includes filters, O-rings, injection components, combustion tube refill materials, and other routine maintenance items to support analyzer uptime and accurate measurements.
  • Cotton swabs, 25 pcs.
  • Furnace injection port, PTFE.
  • Halogen scrubber content refill.
  • Combustion tube refill kit with catalyst and quartz filling.
  • Plastic disposable filter, 5 μm, 10 pcs.
  • Silicone O-ring, 22 x 3 mm, 5 pcs.
  • PTFE O-ring, 6.02 x 2.62 mm, 5 pcs.
  • PTFE stirrer bars, 15 pcs.
  • Furnace injection needle with PTFE sleeve.
  • O-rings for injection and IC port, 5 pcs.
  • PTFE top seal for furnace tube/head.
Catalog No. NC3893355 Supplier Hach Company Supplier No. SMKIT501000
Consumables kit supplying replacement parts for approximately 2,500 injections on the QP1680 TOC/TN analyzer. The kit includes filters, O-rings, injection components, combustion tube refill materials, and other routine maintenance items to support analyzer uptime and accurate measurements.
  • Cotton swabs, 25 pcs.
  • Furnace injection port, PTFE.
  • Halogen scrubber content refill.
  • Combustion tube refill kit with catalyst and quartz filling.
  • Plastic disposable filter, 5 μm, 10 pcs.
  • Silicone O-ring, 22 x 3 mm, 5 pcs.
  • PTFE O-ring, 6.02 x 2.62 mm, 5 pcs.
  • PTFE stirrer bars, 15 pcs.
  • Furnace injection needle with PTFE sleeve.
  • O-rings for injection and IC port, 5 pcs.
  • PTFE top seal for furnace tube/head.