Learn More
Hach Company QP1680 Consumables Kit, 5000 Analyses (for QP1680 TOC/TN Analyzer)
Catalog No. NC2891858 Shop All Hach Company Products
Encompass
Consumables kit for the QP1680 high-temperature TOC/TN laboratory analyzer that provides replacement parts to support approximately 5,000 injections and helps maintain instrument performance and uptime.
  • Provides consumables for approximately 5,000 injections, depending on sample.
  • Consolidates commonly replaced parts into a single maintenance kit.
  • Includes furnace injection ports, injection needles with sleeves, and PTFE top seal.
  • Includes combustion tube and refill kit with catalyst and quartz filling.
  • Includes disposable 5 μm filters and multiple O-rings for furnace and ports.
  • Includes PTFE stirrer bars and cotton swabs for routine maintenance.
  • Helps reduce instrument downtime and simplify scheduled maintenance.
Catalog No. NC2891858 Supplier Hach Company Supplier No. SMKIT501100
Consumables kit for the QP1680 high-temperature TOC/TN laboratory analyzer that provides replacement parts to support approximately 5,000 injections and helps maintain instrument performance and uptime.
  • Provides consumables for approximately 5,000 injections, depending on sample.
  • Consolidates commonly replaced parts into a single maintenance kit.
  • Includes furnace injection ports, injection needles with sleeves, and PTFE top seal.
  • Includes combustion tube and refill kit with catalyst and quartz filling.
  • Includes disposable 5 μm filters and multiple O-rings for furnace and ports.
  • Includes PTFE stirrer bars and cotton swabs for routine maintenance.
  • Helps reduce instrument downtime and simplify scheduled maintenance.