Learn More
Hach Company QP1680 High-Temperature TNb Laboratory Analyser with 65-position autosampler
Catalog No. NC3893354 Shop All Hach Company Products
Encompass
The QP1680 high-temperature laboratory analyser measures total bound nitrogen (TNb) using catalytic combustion and chemiluminescence detection. It includes a 65-position autosampler for 40 mL vials and is engineered for rapid, repeatable laboratory TNb analysis across a wide concentration range.
  • Measures total bound nitrogen (TNb) using chemiluminescence.
  • 65-position autosampler compatible with 40 mL vials.
  • Analysis time approx. 3 minutes per sample.
  • Measuring range TNb 0-30,000 mg/L; LOD 20 μg/L.
  • Sample volume 10-1,000 μL.
  • Catalytic combustion oxidation at 720 °C; furnace max up to 1050 °C.
  • Dimensions 440 x 380 x 700 mm; weight approx. 29 kg.
  • Power supply 100-240 VAC, 50/60 Hz; analyser power up to 750 W.
  • Meets ASTM D8083, EN 12260, ISO 11905-2, NEN-ISO 20236.
Catalog No. NC3893354 Supplier Hach Company Supplier No. LPV448.99.02001
Add to Cart
Add to Cart
Request a Quote
The QP1680 high-temperature laboratory analyser measures total bound nitrogen (TNb) using catalytic combustion and chemiluminescence detection. It includes a 65-position autosampler for 40 mL vials and is engineered for rapid, repeatable laboratory TNb analysis across a wide concentration range.
  • Measures total bound nitrogen (TNb) using chemiluminescence.
  • 65-position autosampler compatible with 40 mL vials.
  • Analysis time approx. 3 minutes per sample.
  • Measuring range TNb 0-30,000 mg/L; LOD 20 μg/L.
  • Sample volume 10-1,000 μL.
  • Catalytic combustion oxidation at 720 °C; furnace max up to 1050 °C.
  • Dimensions 440 x 380 x 700 mm; weight approx. 29 kg.
  • Power supply 100-240 VAC, 50/60 Hz; analyser power up to 750 W.
  • Meets ASTM D8083, EN 12260, ISO 11905-2, NEN-ISO 20236.