Learn More
Enzo Life Sciences AMPIGENE qPCR 1-Step Green Kit Hi-ROX (1000Reactions)
SDP
Supplier:  Enzo Life Sciences ENZNUC1091000
Encompass_Preferred
AMPIGENE qPCR 1-Step Green Kits use the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and qPCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. AMPIGENE qPCR 1-Step Green Kits use a proprietary intercalating dye that does not inhibit PCR. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE qPCR 1-Step Green Kits come with either Lo-ROX or Hi-ROX concentration. For Lo-ROX, see ENZ-NUC108. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below. Applications: PCR.
Catalog No. 50-201-3886