Learn More
Enzo Life Sciences AMPIGENE qPCR 1-Step Probe Kit No-ROX (1000Reactions)
SDP
Catalog No. 502013892
Encompass_Preferred
AMPIGENE qPCR 1-Step Probe Kits use the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and qPCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. AMPIGENE qPCR 1-Step Probe Kits are designed for use on a variety of probes including: Taqma , Molecular Beacons, and Scorpion probes. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE qPCR 1-Step Probe Kits come with either Lo-ROX, Hi-ROX, or No-ROX concentration. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.) Applications: PCR.
Catalog No. 50-201-3892 Supplier Enzo Life Sciences Supplier No. ENZNUC1121000
AMPIGENE qPCR 1-Step Probe Kits use the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and qPCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. AMPIGENE qPCR 1-Step Probe Kits are designed for use on a variety of probes including: Taqma , Molecular Beacons, and Scorpion probes. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE qPCR 1-Step Probe Kits come with either Lo-ROX, Hi-ROX, or No-ROX concentration. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.) Applications: PCR.