Learn More
Biochain Institute Inc dGTP, 400 uL (100 mM)/PK
SDP
Catalog No. 50183029
Encompass_Preferred
BioChain's dNTP reagent combines the highest quality with the most competitive price on the market. The dNTPs are purified with preparative HPLC to achieve at least 99.5% purity.
Catalog No. 50-183-029 Supplier Biochain Institute Inc Supplier No. K6011102400
BioChain's dNTP reagent combines the highest quality with the most competitive price on the market. The dNTPs are purified with preparative HPLC to achieve at least 99.5% purity.