Learn More
Biochain Institute Inc Eva QPCR SuperMix Kit, 400 rxn/PK
SDP
Catalog No. 50183086
Encompass_Preferred
Components in these kits are prepared with pure chemicals according to our proprietary technology. BioChain's QPCR SuperMix is a 2x concentration of premix reagent including Hotstart DNA polymerase and specially formulated real time buffer designed for real-time PCR with intercalator format. Eva and SYBR Green QPCR Supermix is in double-stranded cDNA detection format, while Pro QPCR Supermix is in the format of fluorescent probe-based detection assays, such as TaqMan or molecular beacons.
Catalog No. 50-183-086 Supplier Biochain Institute Inc Supplier No. K5052400
Components in these kits are prepared with pure chemicals according to our proprietary technology. BioChain's QPCR SuperMix is a 2x concentration of premix reagent including Hotstart DNA polymerase and specially formulated real time buffer designed for real-time PCR with intercalator format. Eva and SYBR Green QPCR Supermix is in double-stranded cDNA detection format, while Pro QPCR Supermix is in the format of fluorescent probe-based detection assays, such as TaqMan or molecular beacons.