Learn More
Biochain Institute Inc QCell-Eva One-Step qRT-PCR SuperMix Kit, 200 rxn/PK
SDP
Catalog No. 50183095
Encompass_Preferred
BioChain's QCell-Eva One-Step qRT-PCR SuperMix Kit provides a one-step, simple, robust, inexpensive assay for detection and quantitative analysis of gene expression directly from cells or RNA with intercalator format. QCell-Pro One-Step qRT-PCR SuperMix Kit provides a one-step, simple, robust, inexpensive assay for detection and quantitative analysis of gene expression directly from cells or RNA with probe based format.
Catalog No. 50-183-095 Supplier Biochain Institute Inc Supplier No. K5054200
BioChain's QCell-Eva One-Step qRT-PCR SuperMix Kit provides a one-step, simple, robust, inexpensive assay for detection and quantitative analysis of gene expression directly from cells or RNA with intercalator format. QCell-Pro One-Step qRT-PCR SuperMix Kit provides a one-step, simple, robust, inexpensive assay for detection and quantitative analysis of gene expression directly from cells or RNA with probe based format.