Learn More
Biochain Institute Inc Taq DNA Polymerase, 200 unit/PK
SDP
Catalog No. 50183100
Encompass_Preferred
BioChain's Taq Polymerase polymerizes DNA from a primer annealed to a DNA template in the presence of deoxyribonucleotide triphosphates. It works excellent for not only single PCR but also for multiplex PCR. BioChain's Taq Polymerase is highly purified using a proprietary procedure that virtually eliminates contaminating bacterial DNA and provides excellent reproducibility between lots.
Catalog No. 50-183-100 Supplier Biochain Institute Inc Supplier No. L7051200
BioChain's Taq Polymerase polymerizes DNA from a primer annealed to a DNA template in the presence of deoxyribonucleotide triphosphates. It works excellent for not only single PCR but also for multiplex PCR. BioChain's Taq Polymerase is highly purified using a proprietary procedure that virtually eliminates contaminating bacterial DNA and provides excellent reproducibility between lots.