Learn More
Teknova qPCR DNase Buffer. 100mL, Sterile. 10 pack.
SDP
Catalog No. 502387815
Encompass
10.31mM Magnesium Chloride,10.31mM Calcium Chloride,10.31mM Tris-HCl
Catalog No. 50-238-7815 Supplier Teknova Supplier No. 2P035010PK
Add to Cart
Add to Cart
10.31mM Magnesium Chloride,10.31mM Calcium Chloride,10.31mM Tris-HCl