Learn More
Meridian Life Science qPCR Extraction Control Red, 10ML
SDP
Catalog No. 5000856226
Encompass
Molecular IVD companies continue to experience tighter and tighter regulations in regard to incorporating appropriate positive and negative controls into their assays. Meridian’s qPCR Extraction Control Red uses E. coli cells of a known concentration that contain quantitative PCR positive control DNA sequence (with no known homology to any organism, so that it does not interfere with the detection of the sample target DNA). The cells are added to the sample and lysed and extracted with the sample DNA for downstream amplification by qPCR. The detection of qPCR Extraction Control acts as a PCR positive control and confirms the success of the extraction and amplification steps, avoiding the misinterpretation of a false negative result.
Catalog No. 50-008-56226 Supplier Meridian Life Science Supplier No. MDX02610
Molecular IVD companies continue to experience tighter and tighter regulations in regard to incorporating appropriate positive and negative controls into their assays. Meridian’s qPCR Extraction Control Red uses E. coli cells of a known concentration that contain quantitative PCR positive control DNA sequence (with no known homology to any organism, so that it does not interfere with the detection of the sample target DNA). The cells are added to the sample and lysed and extracted with the sample DNA for downstream amplification by qPCR. The detection of qPCR Extraction Control acts as a PCR positive control and confirms the success of the extraction and amplification steps, avoiding the misinterpretation of a false negative result.