Learn More
Takara Bio QPCR HUMAN REFERENCE TOTAL RNA
SDP
Catalog No. NC9423005
Encompass
Normalization control for gene expression arrays and quantitative PCR studies. A mixture of several tissues provides high representation compared to universal reference made from cell lines.
Catalog No. NC9423005 Supplier Takara Bio Supplier No. 636690
Normalization control for gene expression arrays and quantitative PCR studies. A mixture of several tissues provides high representation compared to universal reference made from cell lines.