Learn More
XpressBio QPCR Mycoplasma Detection Kit
SDP
Catalog No. NC2104235
Encompass_Preferred
The XpressMyco qPCR Mycoplasma Detection Kit is a real-time PCR (qPCR) assay for rapid, quantitative detection of mycoplasma contaminants in cell culture. This kit has 25 reactions.
This item is not returnable. View return policy
Catalog No. NC2104235 Supplier XpressBio Supplier No. XMYCQ25
The XpressMyco qPCR Mycoplasma Detection Kit is a real-time PCR (qPCR) assay for rapid, quantitative detection of mycoplasma contaminants in cell culture. This kit has 25 reactions.