Learn More
Alkali Scientific qPCR/PCR Tube Rack with Lid, Assorted Colors, Fits 96 X0 0.2mL Tubes, Strips or Plates, 5/cs
SDP
Catalog No. 502336240
Encompass_Preferred
Accepts 0.2mL PCR tubes individually, strips of 8, 12, or 96 well PCR plates. Freezer safe and autoclavable polypropylene.
Catalog No. 50-233-6240 Supplier Alkali Scientific Supplier No. R96021
Accepts 0.2mL PCR tubes individually, strips of 8, 12, or 96 well PCR plates. Freezer safe and autoclavable polypropylene.