Learn More
MTC Bio qPCR Sealing Film, Optically Clear, Standard Adhesive, Sterile, 10 bags of 10 seals, 100/pk
Catalog No. NC3737109
Encompass_Preferred
PureAmp™ Sealing Films are compatible with all standard PCR plates.Film and foil options include standard adhesive pierceable aluminum, standard adhesive transparent, optically clear standard adhesive, optically clear non-tacky pressure activated adhesive, and low-adhesive EZ-peel, all of which are guaranteed to keep evaporation rates below 5%.• Multiple types for PCR, qPCR, and ELISA• All types are available sterile• Evaporation rates < 5%• 10 inner bags of 10, 100 per pack• Certified RNase, DNase, DNA, & PCR inhibitor free
Catalog No. NC3737109 Supplier MTC Bio Supplier No. P1001QS
PureAmp™ Sealing Films are compatible with all standard PCR plates.Film and foil options include standard adhesive pierceable aluminum, standard adhesive transparent, optically clear standard adhesive, optically clear non-tacky pressure activated adhesive, and low-adhesive EZ-peel, all of which are guaranteed to keep evaporation rates below 5%.• Multiple types for PCR, qPCR, and ELISA• All types are available sterile• Evaporation rates < 5%• 10 inner bags of 10, 100 per pack• Certified RNase, DNase, DNA, & PCR inhibitor free