Learn More
Medchemexpress LLC QPCT Mouse HEK293 50ug
SDP
Catalog No. 5000183156
Encompass_Preferred
Recombinant mouse glutaminyl-peptide cyclotransferase (QPCT) produced in HEK293 cells and supplied as a lyophilized powder (50 μg). The protein carries N-terminal His and Myc tags, corresponds to accession Q9CYK2-1 (residues A36-L362), and has an approximate molecular weight of 41.6 kDa. It is provided for research applications that require biologically active mouse QPCT.
  • Expressed in HEK293 cells to support eukaryotic post-translational modifications.
  • Contains N-terminal His and Myc tags for purification and detection.
  • Supplied as lyophilized powder for long-term storage and convenient reconstitution.
  • Includes residues A36-L362 of the mature isoform for full-length activity.
  • Approximate molecular weight 41.6 kDa, based on sequence.
  • Suitable for biochemical assays, antibody production, and functional studies.
Catalog No. 50-001-83156 Supplier Medchemexpress LLC Supplier No. HYP7167050UG
Recombinant mouse glutaminyl-peptide cyclotransferase (QPCT) produced in HEK293 cells and supplied as a lyophilized powder (50 μg). The protein carries N-terminal His and Myc tags, corresponds to accession Q9CYK2-1 (residues A36-L362), and has an approximate molecular weight of 41.6 kDa. It is provided for research applications that require biologically active mouse QPCT.
  • Expressed in HEK293 cells to support eukaryotic post-translational modifications.
  • Contains N-terminal His and Myc tags for purification and detection.
  • Supplied as lyophilized powder for long-term storage and convenient reconstitution.
  • Includes residues A36-L362 of the mature isoform for full-length activity.
  • Approximate molecular weight 41.6 kDa, based on sequence.
  • Suitable for biochemical assays, antibody production, and functional studies.