Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Qphos Pd G3

Catalog No. 903027250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-302-7250MG 1 g
90-302-71G 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-302-7250MG Supplier Sigma Aldrich Supplier No. 903027250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

QPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. Qphos Pd G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.QPhos Pd G3 has been used as a precatalyst for the arylboration of isoprene with regioselectivity. It may also be used for a variety of aminination and etherifications of aryl halides.Technology Spotlight: G3 and G4 Buchwald PrecatalystsRegioselective Arylboration of Isoprene and Its Derivatives by Pd/Cu Cooperative CatalysisSynthesis of Complex Drug like Molecules by the Use of Highly Functionalized Bench-Stable Organozinc Reagents

Specifications

CAS 2021255-91-2
Molecular Formula C61H60FeNO3PPdS
Linear Formula C61H60FeNO3PPdS
MDL Number MFCD12911997
Quantity 1 g
Synonym QPhos G3 Palladacycle; QPhos Palladacycle
Molecular Weight (g/mol) 1080.44 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.