Learn More
Waters Corp Performance Maintenance Kit, ACQUITY UPLC Sample Manager-FTN
Catalog No. 50791592 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This performance maintenance kit contains essential components designed to keep an instrument operating at peak performance. It includes a syringe, standard needle, inject valve cartridge, and filters.
  • Syringe
  • Standard needle
  • Inject valve cartridge
  • Filters
  • Sticker for documenting maintenance procedures
Catalog No. 50-791-592 Supplier Waters Corp Supplier No. 201000214
Add to Cart
Add to Cart
Request a Quote
This performance maintenance kit contains essential components designed to keep an instrument operating at peak performance. It includes a syringe, standard needle, inject valve cartridge, and filters.
  • Syringe
  • Standard needle
  • Inject valve cartridge
  • Filters
  • Sticker for documenting maintenance procedures