Learn More
Eurofins Discoverx QRFP-26/P518, 100 µg. (92-1194)
Supplier:  Eurofins Discoverx 921194
Encompass
QRFP-26 (P518) is a 26 amino acid RF-amide peptide found in the brain that is a potent agonist of the G protein-coupled receptor 103 (GPR103). Causes aldosterone release, and may thus regulate adrenal function. Stimulates feeding behavior, metabolic rate, and blood pressure. Provided as 100 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9722