Learn More
Aviva Systems Biology Corp QRFPR Antibody - C-terminal region (ARP60496_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60496_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against QRFPR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human QRFPR. Alternate names: AQ27, GPR103, MGC149217, SP9155ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit, Rat, Zebrafish
Catalog No. 50-287-063