Learn More
MSC QRP Qualatex Finger Cots, Latex, Powder-Free, Large, 720 Pack
Catalog No. 5000796616
Encompass
These finger cots are made from 100% natural latex, designed for precision tasks requiring enhanced grip and dexterity. They are powder-free and feature a sheer film for maximum tactile sensitivity.
  • Made of 100% natural latex in rolled style
  • Miracle Grip polymer coating provides an enhanced wet/dry grip
  • Sheer film for maximum dexterity
  • Economical
  • RoHS compliant
  • Suitable for precision assembly, manufacturing inspection, laboratory, printing/photographic operations, and the beauty industry
Catalog No. 50-007-96616 Supplier MSC Supplier No. 18844563
These finger cots are made from 100% natural latex, designed for precision tasks requiring enhanced grip and dexterity. They are powder-free and feature a sheer film for maximum tactile sensitivity.
  • Made of 100% natural latex in rolled style
  • Miracle Grip polymer coating provides an enhanced wet/dry grip
  • Sheer film for maximum dexterity
  • Economical
  • RoHS compliant
  • Suitable for precision assembly, manufacturing inspection, laboratory, printing/photographic operations, and the beauty industry