Learn More
Aviva Systems Biology Corp QRSL1 Antibody - N-terminal region (ARP42511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against QRSL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human QRSL1. Alternate names: DKFZP564C1278, FLJ10989, FLJ12189, FLJ13447, GatAApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-58249