Learn More
Medchemexpress LLC Qs-21-api | 141256-04-4 | 99.7% | 1990.13 g/mol | C92H148O46 | 1 MG
SDP
Catalog No. 5000213319
Encompass_Preferred
QS-21-Api is a purified saponin adjuvant derived from Quillaja saponaria that enhances antigen-specific humoral and cellular immune responses. Supplied as a powder for research use, it is commonly used to boost immunogenicity in vaccine development studies for oncology and infectious diseases.
  • Enhances both humoral and cellular immune responses
  • Suitable for vaccine adjuvant research applications
  • Provided as a stable powder for long-term storage
  • High chemical purity for reproducible experimental results
  • Compatible with common formulation and dosing workflows
Catalog No. 50-002-13319 Supplier Medchemexpress LLC Supplier No. HY1010921MG
QS-21-Api is a purified saponin adjuvant derived from Quillaja saponaria that enhances antigen-specific humoral and cellular immune responses. Supplied as a powder for research use, it is commonly used to boost immunogenicity in vaccine development studies for oncology and infectious diseases.
  • Enhances both humoral and cellular immune responses
  • Suitable for vaccine adjuvant research applications
  • Provided as a stable powder for long-term storage
  • High chemical purity for reproducible experimental results
  • Compatible with common formulation and dosing workflows