Learn More
Invivogen QS-21 VacciGrade™ STIMULON Adjuvant, Potent Vaccine Adjuvant, Sustainable Sourcing, ≥ 96% Purity
SDP
Catalog No. NC3246796
Encompass
QS-21 VacciGrade™ STIMULON is a highly purified, sterile saponin-based vaccine adjuvant that enhances humoral and Th1-type immune responses, crucial for vaccine efficacy. Sourced from cultured plant cells of *Quillaja saponaria*, it differentiates itself with consistent quality and sustainable sourcing compared to conventional bark-extracted QS-21. Its high purity (≥ 96%) and sterility ensure reliable results for preclinical research, promoting effective antigen uptake and presentation in dendritic cells.
Catalog No. NC3246796 Supplier Invivogen Supplier No. VACQS2125
QS-21 VacciGrade™ STIMULON is a highly purified, sterile saponin-based vaccine adjuvant that enhances humoral and Th1-type immune responses, crucial for vaccine efficacy. Sourced from cultured plant cells of *Quillaja saponaria*, it differentiates itself with consistent quality and sustainable sourcing compared to conventional bark-extracted QS-21. Its high purity (≥ 96%) and sterility ensure reliable results for preclinical research, promoting effective antigen uptake and presentation in dendritic cells.