Learn More
Invivogen QS-21 VacciGrade™ 5mg, Saponin-based vaccine adjuvant (from plant cell culture)
SDP
Catalog No. NC3737505
Encompass
InvivoGen provides QS-21​ VacciGrade™, a sterile and potent vaccine adjuvant obtained from plant cell culture of authenticated native Quillaja saponaria trees [1]. This innovative cultured plant cell production of QS-21 (cpcQS-21) offers a uniform quality and a sustainable alternative to conventional QS-21 extracted from the tree bark (beQS-21). Indeed, the limited cultivation of Chilean soapbark trees Quillaja saponaria, coupled to the intricate and costly bark extraction, makes conventional QS-21 (STIMULON® beQS-21) a scarce and expensive adjuvant.
Catalog No. NC3737505 Supplier Invivogen Supplier No. VACQS215
InvivoGen provides QS-21​ VacciGrade™, a sterile and potent vaccine adjuvant obtained from plant cell culture of authenticated native Quillaja saponaria trees [1]. This innovative cultured plant cell production of QS-21 (cpcQS-21) offers a uniform quality and a sustainable alternative to conventional QS-21 extracted from the tree bark (beQS-21). Indeed, the limited cultivation of Chilean soapbark trees Quillaja saponaria, coupled to the intricate and costly bark extraction, makes conventional QS-21 (STIMULON® beQS-21) a scarce and expensive adjuvant.