Learn More
IBI SCIENTIFIC 3P QS-710 Casting System, UVT Tray w/ Fixture
Catalog No. NC0630600
Encompass_Preferred
For casting a 7cm (wide) x 10cm (long) agarose gel. Includes the QS-710 UVT Casting Tray (IB51020) and the QS-710 Casting Fixture (IB51025).Compatible with the QS-710 Quick Screen Horizontal Electrophoresis System (IB51000).
Catalog No. NC0630600 Supplier IBI SCIENTIFIC 3P Supplier No. IB51030
For casting a 7cm (wide) x 10cm (long) agarose gel. Includes the QS-710 UVT Casting Tray (IB51020) and the QS-710 Casting Fixture (IB51025).Compatible with the QS-710 Quick Screen Horizontal Electrophoresis System (IB51000).