Learn More
IBI SCIENTIFIC 3P QS-710 UV Transmittant Gel Casting Tray
Catalog No. NC9233088
Encompass_Preferred
The QS-710 UV Transmittant Casting Tray mates with the QS-710 Casting Fixture (IB51025) to mold a 7x10cm agarose gel, compatible with the IB51000 QS-710 Quick Screen Horizontal Electrophoresis System. Constructed of durable UVT cast acrylic
Catalog No. NC9233088 Supplier IBI SCIENTIFIC 3P Supplier No. IB51020
The QS-710 UV Transmittant Casting Tray mates with the QS-710 Casting Fixture (IB51025) to mold a 7x10cm agarose gel, compatible with the IB51000 QS-710 Quick Screen Horizontal Electrophoresis System. Constructed of durable UVT cast acrylic