Learn More
TARGETMOL CHEMICALS INC QS11 10MG
SDP
Catalog No. 502579999
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QS11 is a GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1) inhibitor. Modulates ARF-GTP levels and synergizes with the Wnt/(beta)-catenin signaling pathway to upregulate (beta)-catenin nuclear translocation. Also reduces in vitro migration of metastatic human breast cancer cells. purity: 98%
Catalog No. 50-257-9999 Supplier TARGETMOL CHEMICALS INC Supplier No. T402210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QS11 is a GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1) inhibitor. Modulates ARF-GTP levels and synergizes with the Wnt/(beta)-catenin signaling pathway to upregulate (beta)-catenin nuclear translocation. Also reduces in vitro migration of metastatic human breast cancer cells. purity: 98%