Learn More
Medchemexpress LLC HY-12762 5mg Medchemexpress, QS11 CAS:944328-88-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY127625MG
Encompass_Preferred
Medchemexpress, HY-12762 5mg QS11 CAS:944328-88-5 QS11 is an inhibitor of ARFGAP1 (ADP-ribosylation factor GTPase-activating protein 1), with an EC50 of 1.5 µM. QS11 modulates Wnt/β-catenin signaling through an effect on protein trafficking. QS11 inhibits migration of ARFGAP overexpressing breast cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1140