Learn More
Andwin Scientific QSCRIPT 1STEP TOUGHMX 500X20UL
SDP
Catalog No. NC0439730
Encompass_Preferred
The qScript XLT One-Step RT-qPCR ToughMix by Quanta Biosciences is a ready-to-use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates with the use of hybridization probe detection chemistries like the TaqMan 5-hydrolysis probes on real-time PCR systems that do not need a passive reference dye. This product contains: reagent description, qScript XLT One-Step RT-qPCR ToughMix (2X), 2X reaction buffer containing dATP, dCTP, dGTP, dTTP, magnesium chloride, qScript XLT reverse transcriptase, RNase inhibitor protein, hot-start DNA polymerase, AccuVue blue qPCR dye, and stabilizers.
Catalog No. NC0439730 Supplier Andwin Scientific Supplier No. 89236668
The qScript XLT One-Step RT-qPCR ToughMix by Quanta Biosciences is a ready-to-use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates with the use of hybridization probe detection chemistries like the TaqMan 5-hydrolysis probes on real-time PCR systems that do not need a passive reference dye. This product contains: reagent description, qScript XLT One-Step RT-qPCR ToughMix (2X), 2X reaction buffer containing dATP, dCTP, dGTP, dTTP, magnesium chloride, qScript XLT reverse transcriptase, RNase inhibitor protein, hot-start DNA polymerase, AccuVue blue qPCR dye, and stabilizers.