Learn More
Andwin Scientific QSCRIPT CDNA SUPERMIX 100RXNS
SDP
Catalog No. NC0621972
Encompass
The qScript cDNA SuperMix gives a sensitive and easy-to-use solution for two-step qRT-PCR. This concentrated mix offers all needed components for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RNase inhibitor protein, qScript reverse transcriptase, and stabilizers. The mix is optimized for reliable cDNA synthesis over a wide dynamic range of input RNA. It can make 100 reactions.
Catalog No. NC0621972 Supplier Andwin Scientific Supplier No. 101414106
The qScript cDNA SuperMix gives a sensitive and easy-to-use solution for two-step qRT-PCR. This concentrated mix offers all needed components for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RNase inhibitor protein, qScript reverse transcriptase, and stabilizers. The mix is optimized for reliable cDNA synthesis over a wide dynamic range of input RNA. It can make 100 reactions.