Learn More
QST and Fragment Chemistry Installation Kit
Catalog No. 4413691
Encompass_Preferred
Change view
Click to view available options
Product Type:
QST and Fragment Chemistry Installation Kit
Catalog No. Product Type
44-136-91 QST and Fragment Chemistry Installation Kit
1 options
Catalog No. 44-136-91 Supplier LIFE TECHNOLOGIES Supplier No. 4413691
Add to Cart
Add to Cart
Request a Quote
GeneScan™ 500 ROX™ Size Standard is designed for sizing DNA fragments in the 35-500 bp range, provides 16 single-stranded ROX™ dye labeled fragments.
GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.
Order Info
1 Kit
TRUSTED_SUSTAINABILITY
Specifications
Product Type QST and Fragment Chemistry Installation Kit