Learn More
Abcam Qstatin, 5MG
Catalog No. 502671199
Encompass_Preferred
MW 293.2 Da, Purity >=98%. A potent, and selective Vibrio QS inhibitor which affects Vibrio harveyi LuxR homologues, the well-conserved master transcriptional regulators for QS in Vibrio species. QStatin binds tightly to a putative ligand-binding pocket in SmcR, the LuxR homologue in V. vulnificus, and changes the flexibility of the protein, thereby altering its transcription regulatory activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1199 Supplier Abcam Supplier No. AB2871135MG
MW 293.2 Da, Purity >=98%. A potent, and selective Vibrio QS inhibitor which affects Vibrio harveyi LuxR homologues, the well-conserved master transcriptional regulators for QS in Vibrio species. QStatin binds tightly to a putative ligand-binding pocket in SmcR, the LuxR homologue in V. vulnificus, and changes the flexibility of the protein, thereby altering its transcription regulatory activity.
The product is subject to the following: Abcam Restricted Use Statement