Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QSY™ 21 Carboxylic Acid, Succinimidyl Ester

Catalog No. Q20132
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q20132 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q20132 Supplier Invitrogen™ Supplier No. Q20132
  • $445.00
    $333.65 / Each of 1

    Save $111.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

QSY™ 21 carboxylic acid, succinimidyl ester

The amine-reactive quencher, QSY™ 21 succinimidyl ester has a broad and intense absorption (∼661 nm maxima) but no fluorescence making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Wet Ice
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye QSY™ 21
Label Type Quencher Dyes
Product Line QSY
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.