Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QSY™ 9 C5-Maleimide

Catalog No. Q30457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q30457 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q30457 Supplier Invitrogen™ Supplier No. Q30457
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

QSY™ 9 C5-maleimide

The thiol-reactive quencher, QSY™ 9 C5-Maleimide has a broad and intense visible absorption (∼560 nm) but no fluorescence making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Maleimide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye QSY™ 9
Label Type Quencher Dyes
Product Line QSY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.