Learn More
BRANSON ULTRASONICS CORPORATION (3P) QT. CASE INDSTRL STRNGTH 12/CS
Catalog No. NC3721534
Encompass_Preferred
Branson Ultrasonics Ultrasonic Industrial Cleaning Solutions, Size: 1 qt., Concentration: 8 to 10%, For Use With: Cutting Oils, Metal Carbon, pH Range: 11 to 12, Recommended Applications: Safety
Catalog No. NC3721534 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 000955114
Branson Ultrasonics Ultrasonic Industrial Cleaning Solutions, Size: 1 qt., Concentration: 8 to 10%, For Use With: Cutting Oils, Metal Carbon, pH Range: 11 to 12, Recommended Applications: Safety