Learn More
Aviva Systems Biology Corp QTRT1 Antibody - middle region (ARP48946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against QTRT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human QTRT1. Alternate names: FP3235, TGT, TGUTApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-59971