Learn More
Aviva Systems Biology Corp QTRT2 Antibody - C-terminal region (ARP48904_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48904_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Qtrtd1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Qtrtd1. Alternate names: 3110012M05Rik, 4930470H18Rik, AI648807, MGC102598ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-387